Orum International Remote Head Air Sampling Systems
Supplier: HARDY DIAGNOSTICS CA
BAS191 BAS192
CA76509-276EA 5928.01 CAD
CA76509-276 CA76509-278
Orum International Remote Head Air Sampling Systems
Samplers Air Samplers
Remote Head System for contact plates or Petri plates with tri-clamp connection. Completely fabricated in AISI 316 stainless steel.
  • Reduced risk of contamination associated with operator activity
  • Aspirating chamber can be manipulated in multiple directions, according to the unidirectional airflow
  • TRI-clamp device simplifies all the operations of cleaning/sterilization
  • Remote Head System is fully sterilizable
Microbial sampling is programmed through the control unit (Trio.Bas™, AIRBIO, or MULTIFLEX not included) which is generally located outsideMore Product Information