OneRack® Tube Racks, Polypropylene (PP)
Supplier: Heathrow Scientific
HS27551A HS27551B HS27551C HS27551D HS27552A HS27552B HS27552C HS27552D
10147-557CS 165.47 CAD
10147-557 10147-559 10147-561 10147-563 10147-565 10147-567 10147-569 10147-571
OneRack® Tube Racks, Polypropylene (PP)
Racks Tube Racks
OneRack® Tube Racks are tough and highly resistant to acids, bases, solvents, and heat.
  • Made out of Polypropylene
  • Have molded coordinates for tube orientation
  • Stable design for use in water baths
  • Flat labeling area for easier identification
  • Unique tear drop shaped openings for better retention of tubes
  • Offers good chemical and heat resistance (0 to 110 °C)
Durable OneRack® Tube Racks are molded in a single, continuous piece so no assembly is required and there are no detachable pieces.