EZLabpure™ Media Bottle, Closed VersaCap, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
11712-FLS 11722-FLS 11732-FLS 11762-FLS 11772-FLS 11782-FLS
76521-900CS 274.88 CAD
76521-900 76521-902 76521-904 76521-906 76521-908 76521-910
EZLabpure™ Media Bottle, Closed VersaCap, Sterile, Foxx Life Sciences
Bottles Plastic Bottles
The sterile EZlabpure™ Media Bottles, Polystyrene (PS) and PETG, have been designed specifically for Life Science researchers to store common solutions such as tissue culture media, serum, and buffers.
  • PP white VersaCap
  • Sloped shoulder for smooth pouring
  • Molded finger grips
The lower height bottle has features that improve the media flow for easy pouring. The narrow bottle shoulder, combined with the molded finger grips, improves the ability to securely grip the bottle with gloved hands.
Certifications: Made with USP Class VI materials.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable