CryoCube® F570n, ULT Upright Freezers, Eppendorf
Supplier: Eppendorf
F571202035 F571203035 F571402035 F571403035
CA76534-708EA 27104.08 CAD
CA76534-708 CA76534-710 CA76534-720 CA76534-722 MPCA76534-704 MPCA76534-716
CryoCube® F570n, ULT Upright Freezers, Eppendorf
Freezers Ultra Low Temperature Freezers
The Eppendorf ULT Upright Freezers are known for their high energy efficiency and they combine all the enhanced technology with additional vacuum insulation coverage.
  • Reduced energy consumption with high-efficiency fan, compressor and condenser
  • Easy opening with new ergonomic handle that requires less force
  • Easy access to samples with new magnetic closures on inner doors
  • Storage up to 40000 vessels
The ultra-low temperature freezers combine maximum sample security with improved functionality. The advancements decrease power consumption and make freezers more comfortable to use.