Autofil® 2 Bottle Top Filtration Devices, Hydrophilic PES Membrane, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
11012-FLS 11022-FLS 11032-FLS 11412-FLS 11422-FLS 11432-FLS 11512-FLS 11522-FLS 11532-FLS 11612-FLS 11622-FLS 11632-FLS 116-11132-FLS 116-21132-FLS 116-31132-FLS 116-71132-FLS 116-81132-FLS 116-91132-FLS
76521-864CS 300.88 CAD
76521-864 76521-866 76521-868 76521-870 76521-872 76521-874 76521-876 76521-878 76521-880 76521-882 76521-884 76521-886 76521-888 76521-890 76521-892 76521-894 76521-896 76521-898
Autofil® 2 Bottle Top Filtration Devices, Hydrophilic PES Membrane, Sterile, Foxx Life Sciences
Filters Bottle Top Filters
The Autofil® 2 features 0.1, 0.2, or 0.45 μm PES membranes. Perfect for filtering FBS and other viscous solutions. The membrane is low protein binding and surfactant-free.
  • Funnel is compatible with GL 45 thread bottles
  • Units include adapter for connecting to vacuum hose
  • Design includes the NEW EZlabpure™ ergonomic bottle shape with molded finger grip
  • GL 45 caps provides a convenient gripping surface with a frosted area for labeling
Autofil® 2 is available in 250, 500, or 1000 ml complete units with the NEW EZlabpure™ media bottle or funnel only bottle-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable