Human Recombinant Lilra6 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
29813-H02H-100 29813-H02H-1
103886-822EA 555.34 CAD
103886-822 103887-174 CA76477-222 CA76477-574
Human Recombinant Lilra6 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human LILRA6 (Q6PI73-1) (Met1-Asn447) was expressed with the Fc region of human IgG1 at the C-terminus.