Mouse Recombinant CLEC3A Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
50704-M08B-20 50704-M08B-100
103887-262EA 220.22 CAD
103887-262 CA76477-662 CA76477-712 103887-312
Mouse Recombinant CLEC3A Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the mouse CLEC3A (NP_001007224.1) (Met1-Pro196) was expressed with a polyhistidine tag at the C-terminus.