Human Recombinant IFNGR1 Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10338-H02H-100 10338-H02H-500
CA76476-764EA 510.56 CAD
CA76476-764 103886-594 CA76477-966 103887-566
Human Recombinant IFNGR1 Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human IFNGR1 (NP_000407.1) (Met1-Gly245) was expressed with the Fc region of human IgG1 at the C-terminus.