Reusable Taffeta Cleanroom Frocks, Uniform Technology
Supplier: Uniform Technology
CFR-60WH-XS CFR-60WH-S CFR-60WH-M CFR-60WH-L CFR-60WH-XL CFR-60WH-2XL CFR-60WH-3XL CFR-60WH-4XL CFR-60WH-5XL CFR-60NV-XS CFR-60NV-S CFR-60NV-M CFR-60NV-L CFR-60NV-XL CFR-60NV-2XL CFR-60NV-3XL CFR-60NV-4XL CFR-60NV-5XL
76516-302EA 41.03 CAD
76516-302 76516-304 76516-306 76516-308 76516-310 76516-312 76516-314 76516-316 76516-318 76516-320 76516-322 76516-324 76516-326 76516-328 76516-330 76516-332 76516-334 76516-336
Reusable Taffeta Cleanroom Frocks, Uniform Technology
Workwear
Reusable cleanroom frocks are manufactured from 100% polyester Taffeta.
  • Raglan sleeves
  • Military collar
  • Snap front
  • Snap wrists