Reusable ESD Sheer V-Neck Labcoats, Short, ESD Knit Wrists, Uniform Technology
Supplier: Uniform Technology
BR8-42NB-XS BR8-42NB-S BR8-42NB-M BR8-42NB-L BR8-42NB-XL BR8-42NB-2XL BR8-42NB-3XL BR8-42NB-4XL BR8-42NB-5XL
76516-282EA 54.3 CAD
76516-282 76516-284 76516-286 76516-288 76516-290 76516-292 76516-294 76516-296 76516-298
Reusable ESD Sheer V-Neck Labcoats, Short, ESD Knit Wrists, Uniform Technology
Workwear
Reusable ESD labcoats are manufactured from 91% polyester/9% carbon.
  • V-neck design
  • Snap front
  • 2 waist pockets, 1 chest pocket
  • Snap wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.