Reusable ESD StatMaster Labcoats, Long, ESD Knit Wrists, Uniform Technology
Supplier: Uniform Technology
BR5C-47NB-XS BR5C-47NB-S BR5C-47NB-M BR5C-47NB-L BR5C-47NB-XL BR5C-47NB-2XL BR5C-47NB-3XL BR5C-47NB-4XL BR5C-47NB-5XL
76516-246EA 69.08 CAD
76516-246 76516-248 76516-250 76516-252 76516-254 76516-256 76516-258 76516-260 76516-262
Reusable ESD StatMaster Labcoats, Long, ESD Knit Wrists, Uniform Technology
Workwear
Reusable ESD labcoats are manufactured from 93% polyester/7% carbon.
  • Lapel collar
  • Snap front
  • 2 waist pockets, 1 chest pocket
  • ESD knit wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.