Reusable ESD StatStar Labcoats, Long, ESD Knit Wrists, Uniform Technology
Supplier: Uniform Technology
BR51C-44RB-XS BR51C-44RB-S BR51C-44RB-M BR51C-44RB-L BR51C-44RB-XL BR51C-44RB-2XL BR51C-44RB-3XL BR51C-44RB-4XL BR51C-44RB-5XL
76516-120EA 73.08 CAD
76516-120 76516-122 76516-124 76516-126 76516-128 76516-130 76516-132 76516-134 76516-136
Reusable ESD StatStar Labcoats, Long, ESD Knit Wrists, Uniform Technology
Workwear
Reusable ESD labcoats are manufactured from 87% polyester/13% carbon.
  • Lapel collar
  • Snap front
  • 2 waist pockets, 1 chest pocket
  • ESD knit wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.