Reusable ESD StatStar Labcoats, Short, ESD Knit Wrists, Uniform Technology
Supplier: Uniform Technology
BR49AC-44RB-XS BR49AC-44RB-S BR49AC-44RB-M BR49AC-44RB-L BR49AC-44RB-XL BR49AC-44RB2XL BR49AC-44RB3XL BR49AC-44RB4XL BR49AC-44RB5XL
76516-102EA 64.81 CAD
76516-102 76516-104 76516-106 76516-108 76516-110 76516-112 76516-114 76516-116 76516-118
Reusable ESD StatStar Labcoats, Short, ESD Knit Wrists, Uniform Technology
Workwear
Reusable ESD labcoats are manufactured from 87% polyester/13% carbon.
  • Lapel collar
  • Snap front
  • 2 waist pockets, 1 chest pocket
  • ESD knit wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.