Wheat Germ Agglutinin (WGA) Conjugates, Biotium
Supplier: Biotium
29021 29021-1 29022 29022-1 29023 29023-1 29024 29024-1 29025 29025-1 29026 29026-1 29027 29027-1 29029 29059 29029-1 29059-1 29028 29028-1 29073
97065-716EA 615.74 CAD
97065-716 97065-718 97065-720 97065-722 97065-724 97065-726 97065-728 97065-730 97065-732 97065-734 97065-736 97065-738 89410-762 89410-826 10001-624 10001-630 10001-768 10001-792 10018-150 10018-360 75845-664
Wheat Germ Agglutinin (WGA) Conjugates, Biotium
Protein Linking and Labeling Systems
A wheat germ agglutinin (WGA) lectin labeled with Biotium’s CF® Dyes or HRP. It is a carbohydrate-binding lectin that has high affinity for sialic acid and N-acetylglucosamine moieties of glycoproteins. As a result, WGA conjugates will label yeast bud scars and the cell membrane of gram-positive bacteria and mammalian cells.
  • Stain yeast bud scars as well as the cell membrane of gram-positive bacteria and mammalian cells or tissues
  • Suitable for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications
  • A retrograde or anterograde neuronal tracer
  • Withstands fixation and permeabilization
WGA is commonly used to label glycoproteins for imaging of the plasma membrane in live or fixed cells, for staining of tissue sections or for western blotting. WGA can be used as a gram stain to fluorescently label gram-positive bacteria but not gram- bacteria. WGA also binds to the bud scars on budding yeast such as Saccharomyces cerevisiae.
Find the Right Stain for Your Application.
WGA and other lectins are carbohydrate binding proteins that recognize specific sugar moieties on glycoproteins. The presence and distribution of these targets vary between cell types and tissues. As a result, other cell surface stains or other lectin conjugates, Concanavalin A (Con A) conjugates and PNA lectin conjugates, may produce better surface staining and may be more appropriate for your cell type. Lectin conjugates can be used to selectively stain the cell surface of live cells and withstand fixation and permeabilization. When cells are fixed and permeabilized before staining, fluorescent lectins stain both cell surface and organelles in the secretory pathway. Lectins may be toxic or stimulatory to live cells depending on cell type.