Cole-Parmer® Essentials Square Wide-Mouth Plastic Bottles, PPCO, Antylia Scientific
Supplier: Antyila Scientific
06022-62 06022-63 06022-64 06022-65 06022-95
76533-994PK 48.42 CAD
76533-994 76533-996 76533-998 76534-000 76534-002 76534-004
Cole-Parmer® Essentials Square Wide-Mouth Plastic Bottles, PPCO, Antylia Scientific
Bottles Plastic Bottles
Fill and remove samples with ease.
  • Autoclavable
135° C. The wide mouth is easy to fill, store, and transport dry chemicals and environmental samples. Square shape offers visible label surface so bottles are ideal as sample mailers.