Cole-Parmer® Essentials Square Wide-Mouth Plastic Bottles, HDPE, Antylia Scientific
Supplier: Antyila Scientific
06022-01 06022-02 06022-03 06022-04 06022-05 06022-97
76533-984PK 52.13 CAD
76533-984 76533-986 76533-988 76533-990 76533-992 76534-006
Cole-Parmer® Essentials Square Wide-Mouth Plastic Bottles, HDPE, Antylia Scientific
Bottles Plastic Bottles
Maximize sample storage space.
  • Space-saving design eliminates wasted shelf space
  • Ideal for mailing
  • Leakproof polypropylene screw enclosures
These space-saving, leakproof bottles are ideal as sample mailers. Wide mouth is easy to fill. Shape offers visible label surface. All bottles have leakproof polypropylene screw closures. Bottles are not autoclavable.