Cole-Parmer® Essentials Fluorinated HDPE Wide-Mouth Plastic Bottles, Antylia Scientific
Supplier: Antyila Scientific
62151-00 62151-10 62151-20 62151-30 62503-00
76534-338PK 83.29 CAD
76534-338 76534-340 76534-342 76534-344 76534-388
Cole-Parmer® Essentials Fluorinated HDPE Wide-Mouth Plastic Bottles, Antylia Scientific
Bottles Plastic Bottles
Stop worrying about splitting and puncturing.
  • Improved barrier for reduced solvent absorption and penetration
  • Leakproof FLPP closures
  • Stands up to many solvents
FLPP screw closures. Split- and puncture-resistant. Bottles are not autoclavable.