Cole-Parmer® Essentials Wide-Mouth Plastic Bottles, HDPE, Antylia Scientific
Supplier: Antyila Scientific
06047-10 06047-20 06047-30 06047-40 06047-50 06047-60 06055-10
76534-198PK 41.42 CAD
76534-198 76534-200 76534-202 76534-204 76534-206 76534-208 76534-222
Cole-Parmer® Essentials Wide-Mouth Plastic Bottles, HDPE, Antylia Scientific
Bottles Plastic Bottles
Lightweight and flexible for general-purpose laboratory applications.
  • Withstands freezer storage down to −100 °C
  • Translucent and more rigid than LDPE
  • Uniform walls resist splitting and punctures
  • Meets USP Class VI requirements
Suitable for packing, storage, and shipping liquids. All bottles have leakproof polypropylene screw caps. Bottles are not autoclavable.