Cole-Parmer® Essentials Amber Narrow-Mouth Plastic Bottles, HDPE, Antylia Scientific
Supplier: Antyila Scientific
06026-10 06026-20 06026-40 06026-50 06026-60 06026-70
76534-014PK 42.17 CAD
76534-014 76534-016 76534-018 76534-020 76534-022 76534-024
Cole-Parmer® Essentials Amber Narrow-Mouth Plastic Bottles, HDPE, Antylia Scientific
Bottles Plastic Bottles
Bottles meet USP requirements for storage of light-sensitive materials.
  • Reduce UV light transmission
  • Withstands freezer storage down to −100 °C
  • Will not leak with PP screw caps
  • Meets USP Class VI requirements
These amber bottles reduce UV light transmission and meet USP Class VI requirements for storage of light-sensitive materials. They are useful for packaging, shipping, and storage. Leakproof polypropylene screw caps included.Bottles are not autoclavable.