Cole-Parmer® Essentials Plastic Jars, Wide-Mouth, HDPE, Antylia Scientific
Supplier: Antyila Scientific
06046-10 06046-20
76534-194EA 133.99 CAD
76534-194 76534-196
Cole-Parmer® Essentials Plastic Jars, Wide-Mouth, HDPE, Antylia Scientific
Jars General Use Jars
Get better access to samples with the wide-mouth design.
  • Ideal for powder sampling
  • 3.46" (88 mm) neck I.D.
Useful for storing dry chemicals. Jars are made of high-density polyethylene (HDPE) and include high-density polyethylene screw caps. Jars are not autoclavable.