Cole-Parmer® Essentials Jerrican with Handle, HDPE, Antylia Scientific
Supplier: Antyila Scientific
06068-30 06068-31 06068-32
76534-256EA 155.04 CAD
76534-256 76534-258 76534-260
Cole-Parmer® Essentials Jerrican with Handle, HDPE, Antylia Scientific
Bottles Jerricans
Better stability with a wider footprint.
  • Integral spout allows accurate pouring
  • Leak-proof screw closure
  • Milky white polypropylene caps
  • 1.53" (38.9 mm) neck I.D.
Rugged jerricans are built wide and low for better stability. Ideal for transporting and carrying a variety of liquids. Jerricans are not autoclavable.
Certifications: USP Class VI certified