Cole-Parmer® Essentials Carboys with Handles, Heavy-Duty Vacuum, Antylia Scientific
Supplier: Antyila Scientific
06257-50 06257-60
76534-318PK 367.06 CAD
76534-318 76534-320
Cole-Parmer® Essentials Carboys with Handles, Heavy-Duty Vacuum, Antylia Scientific
Bottles Carboys
Withstands temperature applications up to 135 °C (275 °F).
  • No leaks with polypropylene leakproof cap
  • Built-in shoulder handles for easy carrying
  • Shatterproof construction
  • Autoclavable
Designed for large-volume liquid storage and dispensing, these heavy-duty vacuum carboys are compatible with most acids, bases, alcohols, aldehydes, esters and aliphatic hydrocarbons. Constructed from high-quality, laboratory-grade polypropylene resin material that is shatter-proof. These carboys will save money compared to stainless steel and fiberglass-reinforced containers.
Certifications: Meets ISO 13485:2016 and ISO 9001:2015
USP Class VI certified