Cole-Parmer® Essentials Carboy with Spigot and Handles, PP, FDA-Compliant, Antylia Scientific
Supplier: Antyila Scientific
06064-50 06064-60 06064-70
76534-240EA 295.22 CAD
76534-240 76534-242 76534-244
Cole-Parmer® Essentials Carboy with Spigot and Handles, PP, FDA-Compliant, Antylia Scientific
Bottles Carboys
Get a better grip with two-handle construction.
  • Spigot quickly dispenses solutions
  • Leakproof polypropylene cap
  • Easy to read, molded graduations
  • Autoclavable at 250 °F (121 °C)
  • 15.9 mm tubing I.D.
  • 5000 ml graduation intervals
These rugged, durable plastic carboys will meet large volume liquid storage and dispensing needs. Integrated wide handles make them easy to carry and pour, even with gloves on. The attached spigot allows for simple and accurate dispensing of solutions. Round design maximizes mixing efficiency. Polypropylene cap with integrated seal ring ensures leakproof seal. Carboys are chemical resistant.