Cole-Parmer® Essentials Carboy with Spigot and Handles, LDPE, Antylia Scientific
Supplier: Antyila Scientific
06065-22 06065-32 06065-62
76534-246EA 253.07 CAD
76534-246 76534-248 76534-250
Cole-Parmer® Essentials Carboy with Spigot and Handles, LDPE, Antylia Scientific
Bottles Carboys
Easily transport solutions with shoulder handles.
  • Spigot quickly dispenses solutions
  • Leakproof polypropylene cap
  • Easy to read, molded graduations
  • 15.9 mm tubing I.D.
  • 5000 ml graduation intervals
These rugged, durable plastic carboys will meet large volume liquid storage and dispensing needs. Integrated wide handles make them easy to carry and pour, even with gloves on. The attached spigot allows for simple and accurate dispensing of solutions. Round design maximizes mixing efficiency. Polypropylene cap with integrated seal ring ensures leakproof seal. The carboys maintain flexibility and shatterproof safety down to –103 °F ( –75 °C). Carboys are not autoclavable.