Cole-Parmer® Essentials Safety Wash Bottles, LDPE, Antylia Scientific
Supplier: Antyila Scientific
06256-00 06256-10 06256-14 06256-20 06256-50 06256-60 06256-70 06256-81 06256-90
76534-290PK 86 CAD
76534-290 76534-292 76534-294 76534-296 76534-298 76534-300 76534-302 76534-304 76534-306
Cole-Parmer® Essentials Safety Wash Bottles, LDPE, Antylia Scientific
Bottles Wash Bottles
Keep solutions close at hand with easy identification.
  • Quickly identify contents by large lettering
  • Will not leak
  • Polyallomer cap and nozzle assembly
The large lettering on the bottles and color-coded caps allow quick identification of contents. Bottles include a leakproof 43 mm polyallomer cap and nozzle assembly. Please note, these bottle are not recommended for long-term storage of toluene. Bottles are not autoclavable.
Order Now
SPECIFICATIONS
Environmentally Preferable