Cole-Parmer® Essentials Safety Wash Bottles, LDPE, Vented, Antylia Scientific
Supplier: Antyila Scientific
62300-06 62300-16 62300-26 62300-27 62300-28 62300-29 62300-30 62300-36 62300-37 62300-46 62300-47 62300-56 62303-90
76534-354PK 96.23 CAD
76534-354 76534-356 76534-358 76534-360 76534-362 76534-364 76534-366 76534-368 76534-370 76534-372 76534-374 76534-376 76534-378 76534-380
Cole-Parmer® Essentials Safety Wash Bottles, LDPE, Vented, Antylia Scientific
Bottles Wash Bottles
Built-in vent releases built-up pressure, prevents leaking.
  • Silk-screened labels resist chemicals, won’t wear off
  • Easy identification with color-coded closures
These self-venting wash bottles display the chemical name, formula, NFPS hazard rating, DOT symbols, and the Chemical Abstract Service (CAS). Valve mechanism automatically vents volatile solvent vapor and eliminates potentially hazardous problems of solvent drip and sputter. Angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow. A wide mouth makes bottles easy to fill. View contents through translucent bottles and easily identify contents by color-coded closures. The closure and stem are molded as one piece to make them leak-proof. Bottles are not autoclavable.