Cole-Parmer® Essentials Plastic Bottles, Wide-Mouth, PP, Antylia Scientific
Supplier: Antyila Scientific
06057-10 06057-20
76534-230EA 59.75 CAD
76534-230 76534-232
Cole-Parmer® Essentials Plastic Bottles, Wide-Mouth, PP, Antylia Scientific
Bottles Plastic Bottles
Wide-mouth opening provides easier filling and sampling.
  • Leakproof and chemical resistance
  • Free of heavy metals and RoHS compliant
  • BPA and Phthalate free
  • Approved for use from –40 to 250 °F ( –40 to 121 °C)
  • Autoclavable
Manufactured from USP Class VI virgin polypropylene resin, these wide mouth bottles offer excellent chemical resistance and clarity. Precision molded for consistent wall thickness and smooth interior and exterior sufaces. The wide mouth opening is perfect for sample collection and allows easy filling and emptying of solids, powders, and viscous liquids.
Certifications: FDA 21 CFR compliant for food contact.