Reusable ESD Microdenier Cleanroom Sitewear Tops, Uniform Technology
Supplier: Uniform Technology
HSCTM3-48NV-XS HSCTM3-48NV-S HSCTM3-48NV-M HSCTM3-48NV-L HSCTM3-48NV-XL HSCTM3-48NV2XL HSCTM3-48NV3XL HSCTM3-48NV4XL HSCTM3-48NV5XL
76515-980EA 42.49 CAD
76515-980 76515-982 76515-984 76515-986 76515-988 76515-990 76515-992 76515-994 76515-996
Reusable ESD Microdenier Cleanroom Sitewear Tops, Uniform Technology
Workwear
Reusable ESD sitewear tops are manufactured from 99% polyester/1% carbon fabric.
  • Raglan sleeves
  • 3-snap front closure
  • ESD knit cuffs
Sitewear tops feature slits at the bottom of side seams.
Order Now
SPECIFICATIONS
Environmentally Preferable
Garment application Cleanroom Cleanroom
Risk area ESD ESD
Gender Unisex Unisex
Sleeve or leg length Long
Fabric 99% polyester/1% carbon 99% polyester/1% carbon