Reusable Maxima ESD Cleanroom Frocks with Zipper Closure, Uniform Technology
Supplier: Uniform Technology
CFRZC-68WH-XS CFRZC-68WH-S CFRZC-68WH-M CFRZC-68WH-L CFRZC-68WH-XL CFRZC-68WH-2XL CFRZC-68WH-3XL CFRZC-68WH-4XL
76515-868EA 61.71 CAD
76515-868 76515-870 76515-872 76515-874 76515-876 76515-878 76515-880 76515-882
Reusable Maxima ESD Cleanroom Frocks with Zipper Closure, Uniform Technology
Workwear
Cleanroom frocks are manufactured from 99% polyester/1% carbon fabric.
  • Military collar
  • Raglan sleeves
  • ESD knit wrists
  • Coil zipper with top snaps
Suitable for ISO Class 4 or higher cleanrooms.