VHB™ 3M™ GPH-060GF Tapes, TapeCase
Supplier: TAPECASE LTD. TE
8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76524-506PK 45.41 CAD
76524-506 76524-508 76524-510 76524-512 76524-514 76524-516 76524-518 76524-520 76524-522 76524-524 76524-526 76524-528 76524-530 76524-532 76524-534 76524-536 76524-538 76524-540 76524-542 76524-544 76524-546 76524-548 76524-550 76524-552 76524-554 76524-556 76524-558 76524-560 76524-562 76524-564 76524-566 76524-568 76524-570 76524-572 76524-574 76524-576
VHB™ 3M™ GPH-060GF Tapes, TapeCase
Tapes
VHB™ tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
  • Gray, 25 mil, 0.025" (0.6 mm) thick, acrylic adhesive with a conformable, acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives in appropriate applications
  • Maximum temperature: 450 °F (232 °C)
  • Adhesive strength: 14 lbs./inch
VHB™ tape GPH-060GF is double-sided acrylic foam tape with a red polyethylene film liner. The conformable foam provides good contact between substrates even when they are slightly mismatched.