VHB™ 3M™ 4945 Tapes, TapeCase
Supplier: TAPECASE LTD. TE
8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76523-674RL 185.74 CAD
76523-674 76523-676 76523-678 76523-680
VHB™ 3M™ 4945 Tapes, TapeCase
Tapes
VHB™ tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
  • White, 45 mil, 0.045" (1.1 mm), multi-purpose adhesive and firm acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
  • Maximum temperature: 300 °F (149 °C)
  • Adhesive strength: 25 lbs./inch
VHB™ tape 4945 has adhesive on both sides of a firm, foam core with a densified kraft paper liner. It also offers design flexibilityMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable