Argos Technologies® Reusable Inoculating Needles, Antylia Scientific
Supplier: Antyila Scientific
04396-47 04396-48 04396-49 04396-50
76534-552EA 55.24 CAD
76534-552 76534-554 76534-556 76534-558
Argos Technologies® Reusable Inoculating Needles, Antylia Scientific
Spreaders Inoculating Loops and Needles
Nichrome wire composition cools quickly and is less expensive than platinum.
  • Select, inoculate, or transfer microbiological samples
Needles are made from high quality nichrome wire that cools rapidly after sterilization. Available with or without handle and straight or angled needle.