Argos Technologies® Reusable Inoculating Loops without Handles, Antylia Scientific
Supplier: Argos Technologies
04396-35 04396-36 04396-37 04396-38 04396-39
76534-532PK 535.78 CAD
76534-532 76534-534 76534-536 76534-538 76534-540
Argos Technologies® Reusable Inoculating Loops without Handles, Antylia Scientific
Spreaders Inoculating Loops and Needles
Nichrome wire composition cools quickly and is less expensive than platinum.
  • Select, inoculate, or transfer microbiological samples
Argos brand Inoculating Loops are made from high quality twisted nichrome wire. Loops have been designed to slide along agar surfaces without cutting or tearing.
Argos Inoculating Loops cool rapidly and are less expensive than platinum. Loops are supplied without handles and are available calibrated or non-calibrated.
Ordering information: Brass loop handles are sold separately as models 04396-40 and -41.