Argos Technologies® Reusable Inoculating Loop and Needle Holders, Antylia Scientific
Supplier: Antyila Scientific
04396-40 04396-41
76534-528EA 91.67 CAD
76534-528 76534-530
Argos Technologies® Reusable Inoculating Loop and Needle Holders, Antylia Scientific
Spreaders Inoculating Loops and Needles
Save more with these reusable loop and needle handles.
  • Handles (6 or 8") are sold separately for some loops and needles
Reusable Inoculating Loops with handles are used to select, inoculate or transfer microbiological samples. Handles are insulated to protect the hand during use.
Loops and Handles cool rapidly after sterilization.
Ordering information: Loops and without handles are sold separately as models 04396-35 through -39. Needle without handle is available as model 04396-49.