Cole-Parmer® Essentials Lab Inoculation Needles, Sterile, Polystyrene
Supplier: Antyila Scientific
06231-06 06231-09
76534-522CS 175.7 CAD
76534-522 76534-524 76534-526
Cole-Parmer® Essentials Lab Inoculation Needles, Sterile, Polystyrene
Spreaders Inoculating Loops and Needles
Save time and eliminate cross-contamination.
  • Constructed of durable polystyrene
Smooth and flexible needles are constructed of high-impact polystyrene. Straight needles are ideal for picking single colonies. Color coding identifies inoculation volume easily.
Certifications: Each case is supplied with a quality certificate confirming sterility and accuracy.
Packaging: Packed sterile in tamperproof resealable bags.