Cole-Parmer® Essentials Disposable Inoculation Loops, Antylia Scientific
Supplier: Antyila Scientific
06231-02 06231-03 06231-04 06231-05 06231-07 06231-08
76534-510CS 146.85 CAD
76534-510 76534-512 76534-514 76534-516 76534-518 76534-520
Cole-Parmer® Essentials Disposable Inoculation Loops, Antylia Scientific
Spreaders Inoculating Loops and Needles
Save time and eliminate cross-contamination.
  • Constructed of durable polystyrene
Smooth and flexible loops are constructed of high-impact polystyrene. Loops ensure uniform streaking without damaging your gel surface. Color coding identifies inoculation volume easily.
Certifications: Each case is supplied with a quality certificate confirming sterility and accuracy.
Packaging: Packed sterile in tamperproof resealable bags.