VHB™ 3M™ GPH-110GF Tapes, TapeCase
Supplier: TAPECASE LTD. TE
8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76524-578PK 44.73 CAD
76524-578 76524-580 76524-582 76524-584 76524-586 76524-588 76524-590 76524-592 76524-594 76524-596 76524-598 76524-600 76524-602 76524-604 76524-606 76524-608 76524-610 76524-612 76524-614 76524-616 76524-618 76524-620 76524-622 76524-624 76524-626
VHB™ 3M™ GPH-110GF Tapes, TapeCase
Tapes
VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
  • Gray, 1.1 mm (45 mil), acrylic adhesive with a conformable, acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives in appropriate applications
  • Maximum temperature: 232 °C (450 °F)
  • Adhesive strength: 21 lbs./inch
VHB™ Tape GPH-110GF is a 1.1 mm (0.045") thick, gray double-sided acrylic foam tape with a red polyethyleneMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable