VHB™ 3M™ 4930 Tapes, TapeCase
Supplier: TAPECASE LTD. TE
8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76523-412RL 17.08 CAD
76523-412 76523-414 76523-416 76523-418 76523-420 76523-422 76523-424 76523-426 76523-428 76523-430 76523-432 76523-434 76523-436 76523-438 76523-440 76523-442 76523-444 76523-446 76523-448 76523-450 76523-452 76523-454 76523-456 76523-458 76523-460 76523-462 76523-464 76523-466 76523-468 76523-470 76523-472 76523-474 76523-476 76523-478 76523-480 76523-482 76523-484 76523-486 76523-488 76523-490 76523-492 76523-494 76523-496 76523-498 76523-500 76523-502 76523-504 76523-506 76523-508 76523-510 76523-512 76523-514 76523-516 76523-518 76523-520 76523-522 76523-524 76523-526 76523-528 76523-530 76523-532 76523-534 76523-536 76523-538 76523-540 76523-542 76523-544 76523-546 76523-548 76523-550 76523-552 76523-554 76523-556 76523-558 76523-560 76523-562 76523-564 76523-566 76523-568 76523-570 76523-572 76523-574 76523-576 76523-578 76523-580 76523-582 76523-584 76523-586 76523-588 76523-590 76523-592 76523-594 76523-596 76523-598 76523-600 76523-602 76523-604 76523-606 76523-608 76523-610 76523-612 76523-614 76523-616 76523-618 76523-620 76523-622 76523-624 76523-626 76523-628 76523-630 76523-632 76523-634 76523-636 76523-638 76523-640 76523-642 76523-644 76523-646 76523-648 76523-650 76523-652 76523-654 76523-656 76523-658 76523-660 76523-662 76523-664 76523-666 76523-668 76523-670 76523-672
VHB™ 3M™ 4930 Tapes, TapeCase
Tapes
VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
  • White, 25 mil (0.6 mm), general-purpose adhesive and firm acrylic foam core
  • Permanent bonding provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
  • Maximum temperature: 300 °F (149 °C)
  • Adhesive strength: 20 lbs./inch
VHB™ Tape 4930 is a firm, double-sided, pressure-sensitive, closed-cell acrylic foam tape. High-strength and durable, with a clean appearance.