Cole-Parmer® Cryogenic Vials, External Thread, Sterile, Antylia Scientific
Supplier: Antyila Scientific
78903-21 78903-20
76534-404CS 361.73 CAD
76534-404 76534-406
Cole-Parmer® Cryogenic Vials, External Thread, Sterile, Antylia Scientific
Vials
Stores samples as low as –196 °C.
  • DNase-, RNase- and pyrogen-free
  • Radiation sterilized
  • HDPE cap
These polypropylene cryogenic vials feature a self-standing star base designed for easy one-handed manipulation when used with a support rack.
Vials include an internal U-shaped bottom, large white frosted labeling area, silk screen graduations and a leak-proof HDPE cap.
Ideal for used in vapor phase of liquid nitrogen but are not recommended for use in the liquid phase of liquid nitrogen.