Cole-Parmer® Essentials Wide-Mouth Sample Polypropylene Containers, Antylia Scientific
Supplier: Antyila Scientific
06101-40 06101-60 06101-70 06101-11 06101-50 06101-21 06101-31
76534-484PK 84.13 CAD
76534-484 76534-486 76534-488 76534-490 76534-492 76534-494 76534-496
Cole-Parmer® Essentials Wide-Mouth Sample Polypropylene Containers, Antylia Scientific
Sample Containers General Purpose Containers
Mix and store in one vessel.
  • Easily attach to blenders for in-container mixing
  • Durable for general storage—chemical-resistant polypropylene
  • Autoclavable
Chemical-resistant polypropylene (PP) containers come with white PP linerless lids.
Accessories information: PP lid.