Cole-Parmer® Essentials Cylindrical HDPE Jars, Antylia Scientific
Supplier: Antyila Scientific
63100-14 63100-12 63100-20 63100-10 63100-22 63100-16 63100-18
76534-446PK 105.48 CAD
76534-446 76534-448 76534-450 76534-452 76534-454 76534-456 76534-458
Cole-Parmer® Essentials Cylindrical HDPE Jars, Antylia Scientific
Jars General Use Jars
Get easy sample access.
  • Wide neck for easy powder sampling
  • Include screw caps to keep samples intact
  • Won't break—made of high-density polyethylene for sturdy storage
  • Non-autoclavable
High-density polyethylene (HDPE) containers feature a wide neck which is ideal for powders and solid specimens. Screw-on caps with insert are included.
Accessories information: Screw-on caps with insert.