Cole-Parmer® Essentials Narrow-Mouth LDPE Bottles, Antylia Scientific
Supplier: Antyila Scientific
06038-29 06038-23 06038-21 06038-25 06038-27
76534-460PK 119.94 CAD
76534-460 76534-462 76534-464 76534-466 76534-468
Cole-Parmer® Essentials Narrow-Mouth LDPE Bottles, Antylia Scientific
Bottles Plastic Bottles
Keep samples contained with leakproof design.
  • Leakproof and chemical resistant for storing and handling liquids and solids
  • LDPE bottles feature transparent, food-grade design
  • Non-autoclavable
Transulcent, flexible, low density polyethylene bottles (LDPE) are manufactured with approved FDA compliant resins that are ideal for food product applications. Bottles are complete with leakproof polypropylene (PP) caps.
Accessories information: Leakproof closure.