Cole-Parmer® Essentials HDPE Sample Bottles with Polypropylene Caps, Antylia Scientific
Supplier: Antyila Scientific
06049-00 06049-01 06050-05
76534-424CS 1305.29 CAD
76534-424 76534-426 76534-428
Cole-Parmer® Essentials HDPE Sample Bottles with Polypropylene Caps, Antylia Scientific
Bottles Plastic Bottles
Store samples without breaking your budget.
  • Provide good chemical resistance for a wide range of uses
  • Bottles are made of HDPE; included cap is made of polypropylene and lined with polyethylene foam
  • Non-autoclavable
Bottles feature good chemical resistance and include polypropylene caps with polyethylene foam liner.
Accessories information: Polypropylene caps with polyethylene foam liner.