VWR® Ultra Low Liquid-in-Glass Thermometers
Supplier: VWR International
60760-1311 60760-1511
76491-632CS 1648.81 CAD
76491-632 76491-634
VWR® Ultra Low Liquid-in-Glass Thermometers
Thermometers Analog Thermometers
Partial and total immersion thermometers for low temperature applications.
  • Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Tested and inspected by our USA quality team
Designed for ultra-low temperature use to ensure that the meniscus will be visible in the appropriate low temperature applications, these thermometers feature mercury free liquid against lead-free, white back glass.
Certifications: Traceable to NIST. Compliant with ANSI/SAMA Z236.1-1983.