VWR® Dry Block/Incubator Liquid-in-Glass Thermometer, Non-Toxic Fill
Supplier: VWR International
76491-620 76491-622
76491-620CS 902.94 CAD
76491-620 76491-622
VWR® Dry Block/Incubator Liquid-in-Glass Thermometer, Non-Toxic Fill
Thermometers Analog Thermometers
Designed for use in all standard heating and drying blocks, water baths and incubators.
  • Partial immersion
  • Individual serial numbers
  • Same accuracy as similar mercury thermometers, but without the hazards
Contains enviro safe, non-toxic, non-hazardous, biodegradable green liquid against white back glass.
Certifications: Traceable to NIST; hand inspected in triple-accredited/-registered ISO/IEC 9001:2008, ISO/IEC 14001:2004, ISO/IEC 17025:2005 facility and A2LA Accredited Calibration Laboratory.
Delivery information: Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy indicating accuracy traceable to NIST.