VWR® Environmentally Friendly, Easy Read Thermometers, Individually Calibrated
Supplier: VWR International
60202-0011 60202-0111 60202-0411 60202-0811
76491-584CS 1678.57 CAD
76491-584 76491-586 76491-588 76491-590 76491-592
VWR® Environmentally Friendly, Easy Read Thermometers, Individually Calibrated
Thermometers Analog Thermometers
Each easy to read thermometer includes an individualized thermometer calibration report stating corrections to one-tenth of the smallest division, unique report number, and NIST standards used for calibration.
  • EnviroKleen™ certified
  • Non-toxic, biodegradable, enviro-safe liquid, against lead-free, yellow back glass
  • Provides easy readability and fewer reading errors
  • Individually serialized
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Certifications: Traceable to NIST, individually serialised.