VWR® Probeless Indoor/Outdoor Digital Thermometer
Supplier: VWR
VWR®
60900-2811
76491-652CS 1409.9 CAD
76491-652
VWR® Probeless Indoor/Outdoor Digital Thermometer
Thermometers Digital Thermometers
Designed for use in high moisture or wet environments, this digital thermometer is used in green houses, sample prep areas, water treatment facilities and aquariums.
  • Reset button allows for a new monitoring period
  • Keyhole for mounting
  • Easy to read
  • Water resistant IP 65
Displays the current temperature and registers the maximum and minimum temperatures attained since the last reset.
Delivery information: Includes LR44 battery.