VWR® Incubator Verification Thermometers
Supplier: VWR International
76491-666 76491-668 76491-678
76491-666CS 1047.07 CAD
76491-666 76491-668 76491-678
VWR® Incubator Verification Thermometers
Thermometers Analog Thermometers
These mercury free thermometers are traceable to NIST and cover ranges from 15 to 50 °C.
  • Verify sample temperature in incubators
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Thermometers contain organic liquid; featuring permanently fused markings and lead-free glass
  • PFA coating safely contains glass and liquid in case of breakage
  • Individual serial number
Insert the thermometer into the bottle and place near the samples for best results.
Certifications: Traceable to NIST.
Delivery information: Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability plus VELCRO® and magnetic attachments.