VWR® Blood Bank Verification Thermometers
Supplier: VWR
VWR®
61004-0411
76491-676CS 743.95 CAD
76491-676
VWR® Blood Bank Verification Thermometers
Thermometers Analog Thermometers
These mercury free thermometers are traceable to NIST and cover ranges from −5 to 20 °C.
  • Verify sample temperatures in blood bank refrigerators
  • Thermometers contain organic liquid, or EnviroKleen™ certified Enviro-Safe® liquid; feature permanently fused markings and lead free glass
  • Manufactured and tested by our USA quality team; individually serialized
  • Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
Accurate temperature reading is achieved by using the filled bottle (included), which mimics the sample. Insert the thermometer into the bottle and place near the samples for best results. PFA coating safely contains glass and liquid in case of breakage.
Certifications: Traceable to NIST.
Delivery information: Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability plus VELCRO® and magnetic attachments.