VWR® Incubator Verification Thermometers with Calibration Report
Supplier: VWR International
76491-578 76491-580
76491-578CS 3330.32 CAD
76491-578 76491-580
VWR® Incubator Verification Thermometers with Calibration Report
Thermometers Analog Thermometers
These mercury-free thermometers are individually calibrated at specific Temperature points specifically for use in incubators.
  • Verify sample temperatures in incubators
  • Provides accurate temperature readings without sample contamination
  • Filled bottle (included) mimics the sample
  • Permanently fused markings and lead-free glass
  • Individual serial number
To use, simply screw thermometer with cap into bottle and attach to test sample.
Certifications: Traceable to NIST.
Delivery information: Supplied with individual Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration.